Merck
Evaluation of an antispasmodic agent. Methixene hydrochloride (Trest).
JAMA (1966-03-07)
ABSTRACT
Methixene hydrochloride, an anticholinergic agent, may be useful in the symptomatic treatment of functional bowel disorders. Adverse effects reported from its use are those of the anticholinergic drugs in general.
Product Description
Metixene hydrochloride, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.