Merck
6,7-Bis(bromomethyl)-2,11,18,21,24-pentaoxatetracyclo.
Acta crystallographica. Section C, Crystal structure communications (2001-03-16)
W Sim, J Y Lee, J S Kim, J G Kim, I H Suh
ABSTRACT
The 17-crown-5 unit, C(26)H(26)Br(2)O(5), consisting of a 1,2-bis(bromomethyl) group, three benzo groups and diethylene glycol, was prepared from the reaction of 1,2,4,5-tetrakis(bromomethyl)benzene and bis-phenol in the presence of sodium hydride as a base. This molecule seems to offer an internal cavity for the formation of a host--guest complex.
Sigma-Aldrich
1,2,4,5-Tetrakis(bromomethyl)benzene, 95%