Merck
  • Lithium diisopropylamide-mediated ortholithiation of 2-fluoropyridines: rates, mechanisms, and the role of autocatalysis.
Lithium diisopropylamide-mediated ortholithiation of 2-fluoropyridines: rates, mechanisms, and the role of autocatalysis.
The Journal of organic chemistry (2012-12-29)
Lekha Gupta, Alexander C Hoepker, Yun Ma, Mihai S Viciu, Marc F Faggin, David B Collum
ABSTRACT
Lithium diisopropylamide (LDA)-mediated ortholithiations of 2-fluoropyridine and 2,6-difluoropyridine in tetrahydrofuran at -78 °C were studied using a combination of IR and NMR spectroscopic and computational methods. Rate studies show that a substrate-assisted deaggregation of LDA dimer occurs parallel to an unprecedented tetramer-based pathway. Standard and competitive isotope effects confirm post-rate-limiting proton transfer. Autocatalysis stems from ArLi-catalyzed deaggregation of LDA proceeding via 2:2 LDA-ArLi mixed tetramers. A hypersensitivity of the ortholithiation rates to traces of LiCl derives from LiCl-catalyzed LDA dimer-monomer exchange and a subsequent monomer-based ortholithiation. Fleeting 2:2 LDA-LiCl mixed tetramers are suggested to be key intermediates. The mechanisms of both the uncatalyzed and catalyzed deaggregations are discussed. A general mechanistic paradigm is delineated to explain a number of seemingly disparate LDA-mediated reactions, all of which occur in tetrahydrofuran at -78 °C.
Sigma-Aldrich
2,6-Difluoropyridine, 99%