Merck
  • The supramolecular structure of 6-hydroxy-1,3-benzoxathiol-2-one (tioxolone).
The supramolecular structure of 6-hydroxy-1,3-benzoxathiol-2-one (tioxolone).
Acta crystallographica. Section C, Crystal structure communications (2004-06-05)
Maureen Byres, Philip J Cox
ABSTRACT
The planar molecules of 6-hydroxy-1,3-benzoxathiol-2-one, C(7)H(4)O(3)S, are linked by extensive O-H.O and C-H.O hydrogen bonding and are further stablilized by face-to-face pi-pi interactions.
Sigma-Aldrich
6-Hydroxy-1,3-benzoxathiol-2-one, ≥98%