Merck
  • Reactions of methylenecyclopropanes with phenylsulfenyl chloride and phenylselenyl chloride.
Reactions of methylenecyclopropanes with phenylsulfenyl chloride and phenylselenyl chloride.
The Journal of organic chemistry (2004-04-13)
Le-Ping Liu, Min Shi
ABSTRACT
The reaction of methylenecyclopropanes 1 with phenylsulfenyl chloride or phenylselenyl chloride gives (cyclobut-1-enylsulfanyl)benzene or (cyclobut-1-enylselanyl)benzene along with ring-opened product in good total yields at 0 degrees C in various solvents. A plausible mechanism has been proposed.
Sigma-Aldrich
Phenylselenyl chloride, 98%
Pricing and availability is not currently available.