Merck
  • Anti-polymorphonuclear neutrophil antibodies in patients with leukopenia or neutropenia.
Anti-polymorphonuclear neutrophil antibodies in patients with leukopenia or neutropenia.
International journal of laboratory hematology (2009-07-15)
N E Riera, M Rosso Saltó, V Galán, K Canalejo, M Khoury, M Aixalá, G L Kantor, M Vermeulen, R Bengió, M M E De Bracco
ABSTRACT
Immune humoral neutropenia (Np) could be the consequence of anti-polymorphonuclear neutrophil (PMN) antibodies, circulating immune complexes (CIC) and/or antibodies against myeloid precursors. Granulocyte immunofluorescence test (GIFT) and a leukoagglutination technique (LAGT) assays are recommended for its diagnosis. Fifty adult patients with secondary Np were screened for anti-PMN. GIFT by flow cytometry from viable PMN and LAGT were employed. In addition, CIC levels, low expression of CD16(b) (CD16 (b)(low)), PMN phenotype and sera tumor necrosis factor-alpha (TNF-alpha) were also evaluated. Direct IgG-PMN binding (dir-GIFT) was positive in 16% of the patients. Antibodies against autologous PMN were detected in 32% of the samples by indirect (ind)-GIFT and demonstrated in 70% of the sera by both ind-GIFT and/or LAGT. Predominance of human neutrophil alloantigen (HNA)-1b and HNA-2 expression was confirmed. CD16(b)(low) was detected in 16% of the patient's PMN and TNF-alpha in 68% of sera patients. Our results suggest that diagnosis of immune Np in the laboratory may be improved by focusing on patient's PMN together with the assessment of cellular markers.
Sigma-Aldrich
Globulins Cohn fraction II, III human, lyophilized powder
Pricing and availability is not currently available.
Sigma-Aldrich
Globulins Cohn fraction IV-4 human
Pricing and availability is not currently available.