Merck
  • Comparative effect of pioglitazone, quercetin and hydroxy citric acid on the status of lipid peroxidation and antioxidants in experimental non-alcoholic steatohepatitis.
Comparative effect of pioglitazone, quercetin and hydroxy citric acid on the status of lipid peroxidation and antioxidants in experimental non-alcoholic steatohepatitis.
Journal of physiology and pharmacology : an official journal of the Polish Physiological Society (2014-03-14)
K M Surapaneni, M Jainu
ABSTRACT
Oxidative stress is one of the key mechanisms responsible for liver damage and disease progression in non-alcoholic steatohepatitis (NASH). Antioxidants try to combat the oxidative stress and minimise the oxidative stress. In this study, the comparative effect of pioglitazone, quercetin and hydroxy citric acid on lipid peroxidation and antioxidants in experimentally induced NASH has been studied. The levels of lipid peroxidation products, malondialdehyde (MDA), glutathione (GSH), catalase (CAT), superoxide dismutase (SOD), glutathione peroxidase (GPx), glutathione reductase (GR) and glutathione-S-transferase (GST) were estimated in experimental NASH. The levels of lipid peroxidation products (MDA) have been significantly increased in experimentally induced NASH group compared to the control group. The experimental NASH rats treated with pioglitazone, quercetin and with hydroxy citric acid showed significant reduction in MDA levels when compared with that of NASH induced group. Non enzymatic antioxidant such as GSH and antioxidant enzymes such as catalase, SOD, GPx, GR, and GST were decreased significantly in experimental NASH, compared to that of control group. The experimental NASH rats treated with pioglitazone showed marked increase in the levels of GSH, catalase and SOD but no significant effect on the levels of GPx, GR and GST levels when compared to the experimentally induced NASH group. Experimental NASH rats treated with hydroxy citric acid showed marked increase in the levels of GSH and catalase but no significant effect on the levels of SOD, GPx, GR and GST levels when compared to the experimentally induced NASH group. On contrary to these two drugs, the experimental NASH rats treated with quercetin showed significant increase in the levels of antioxidants as follows: GSH, catalase, SOD, GPx, GR and GST when compared with that of NASH induced group. We concluded that the levels of nonenzymatic and enzymatic antioxidants found to be decreased in experimentally induced NASH. The antioxidant property of quercetin could be more beneficial in treatment of NASH when compared to pioglitazone and hydroxy citric acid.
Sigma-Aldrich
CKI-7 dihydrochloride, ≥98% (HPLC), solid
Pricing and availability is not currently available.