Merck
  • Liver failure after major liver resection: risk assessment by using preoperative Gadoxetic acid-enhanced 3-T MR imaging.
Liver failure after major liver resection: risk assessment by using preoperative Gadoxetic acid-enhanced 3-T MR imaging.
Radiology (2013-08-15)
Andreas Wibmer, Alexander M Prusa, Richard Nolz, Thomas Gruenberger, Martin Schindl, Ahmed Ba-Ssalamah
ABSTRACT
To determine if gadoxetic acid-enhanced magnetic resonance (MR) imaging with measurement of relative liver enhancement (RLE) on hepatobiliary phase images can allow preoperative assessment of the risk of liver failure after major liver resection. The local institutional review committee approved this retrospective analysis and waived written informed consent. The study included 73 patients (39 men; median age, 64.4 years) who underwent gadoxetic acid-enhanced 3-T MR imaging before resection of three or more liver segments. RLE was calculated as the ratio of signal intensity measurements of the liver parenchyma before and 20 minutes after intravenous administration of gadoxetic acid. RLE was assessed in each liver segment and the mean value of all segments was used for analysis. Posthepatectomy liver failure was defined according to the "50-50 criteria" (ie, prothrombin time <50% and serum bilirubin >50 µmol/L on postoperative day 5) and the International Study Group of Liver Surgery (ISGLS) classification. The association of RLE and liver failure was tested with univariate and multivariate logistic regression analysis. In addition to RLE, the latter also included demographic, clinical, operative, and histologic variables. Patients with liver failure according to the 50-50 criteria (n = 3) had significantly lower RLE (54.5%) than those without (125.6%) (P = .009). According to ISGLS criteria, RLE was 112.5% in patients with grade A liver failure (n = 20), 88.4% in patients with grade B (n = 7), 41.7% (n = 2) in patients with grade C, and 136.5% (P < .001) in those without liver failure. In a logistic regression analysis, RLE was inversely related to the probability of liver failure according to the 50-50 (P = .02) and ISGLS (P < .001) criteria. In a multivariate analysis, RLE was independently associated with a higher probability of liver failure according to ISGLS classification (P = .003). Gadoxetic acid-enhanced MR imaging can help with the assessment of the risk for liver failure after major liver resection.
Sigma-Aldrich
Bilirubin, purum, ≥95.0% (UV)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
50 g
1 Estimated to ship on 09 March 2025
$96.80
250 g
1 Estimated to ship on 09 March 2025
$196.00
500 g
1 Estimated to ship on 09 March 2025
$304.00
1 kg
1 Estimated to ship on 09 March 2025
$339.00
Sigma-Aldrich
Bilirubin, ≥98% (EmM/453 = 60), powder
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
50 g
1 Estimated to ship on 09 March 2025
$96.80
250 g
1 Estimated to ship on 09 March 2025
$196.00
500 g
1 Estimated to ship on 09 March 2025
$304.00
1 kg
1 Estimated to ship on 09 March 2025
$339.00