Merck
  • DNA-templated release of functional molecules with an azide-reduction-triggered immolative linker.
DNA-templated release of functional molecules with an azide-reduction-triggered immolative linker.
Chemical communications (Cambridge, England) (2011-03-04)
Katarzyna Gorska, Alex Manicardi, Sofia Barluenga, Nicolas Winssinger
ABSTRACT
Nucleic acid templated reactions have attracted significant attention for nucleic acid sensing. Herein we report a general design which extends the potential of nucleic acid templated reactions to unleash the function of a broad diversity of small molecules such as a transcription factor agonist, a cytotoxic or a fluorophore.
Sigma-Aldrich
4-Aminobenzyl alcohol, 98%
Pricing and availability is not currently available.