Merck
  • Mass spectrometric characterization of phosphorylated peptides using MALDI in-source decay via redox reactions.
Mass spectrometric characterization of phosphorylated peptides using MALDI in-source decay via redox reactions.
Journal of mass spectrometry : JMS (2012-02-24)
Daiki Asakawa, Mitsuo Takayama
ABSTRACT
Matrix-assisted laser desorption/ionization in-source decay (MALDI-ISD) has been used for characterization of a phosphorylated peptides and proteins because labile phosphate group is not lost during the MALDI-ISD process. The conventional MALDI-ISD is initiated by the hydrogen transfer from reducing matrix molecules to peptide backbone, leading to c'- and z'-series ions. In contrast, when an oxidizing chemical 5-nitrosalicylic acid (5-NSA) is served as the MALDI-ISD matrix, a- and x-series ions are specifically generated by hydrogen abstraction from peptide backbone to matrix molecule. The 5-NSA provides useful complementary information to the conventional MALDI-ISD for the analysis of amino acid sequencing and site localization of phosphorylation in peptides. The MALDI-ISD with reducing and oxidizing matrix could be a useful method for the de novo peptide sequencing.
Product Description
Supelco
1,5-Diaminonaphthalene, matrix substance for MALDI-MS, ≥99.0% (HPLC)