Merck
  • Cu- and Pd-catalyzed asymmetric one-pot tandem addition-cyclization reaction of 2-(2',3'-alkadienyl)-beta-keto esters, organic halides, and dibenzyl azodicarboxylate: an effective protocol for the enantioselective synthesis of pyrazolidine derivatives.
Cu- and Pd-catalyzed asymmetric one-pot tandem addition-cyclization reaction of 2-(2',3'-alkadienyl)-beta-keto esters, organic halides, and dibenzyl azodicarboxylate: an effective protocol for the enantioselective synthesis of pyrazolidine derivatives.
Organic letters (2004-06-18)
Shengming Ma, Ning Jiao, Zilong Zheng, Zhichao Ma, Zhan Lu, Longwu Ye, Youqian Deng, Guofei Chen
ABSTRACT
[reaction: see text] Optically active pyrazolidine derivatives have been constructed by the Cu- and Pd-catalyzed asymmetric one-pot tandem addition-cyclization reaction of 2-(2',3'-dienyl)-beta-ketoesters, organic halides, and dibenzyl azodicarboxylate. The absolute configurations of the final products depend on the structure of the ligand.
Sigma-Aldrich
Dibenzyl azodicarboxylate, technical grade, 90%