Merck
Genetic basis of hyperlysinemia.
Orphanet journal of rare diseases (2013-04-11)
Sander M Houten, Heleen Te Brinke, Simone Denis, Jos Pn Ruiter, Alida C Knegt, Johannis Bc de Klerk, Persephone Augoustides-Savvopoulou, Johannes Häberle, Matthias R Baumgartner, Turgay Coşkun, Johannes Zschocke, Jörn Oliver Sass, Bwee Tien Poll-The, Ronald Ja Wanders, Marinus Duran
ABSTRACT
Hyperlysinemia is an autosomal recessive inborn error of L-lysine degradation. To date only one causal mutation in the AASS gene encoding α-aminoadipic semialdehyde synthase has been reported. We aimed to better define the genetic basis of hyperlysinemia. We collected the clinical, biochemical and molecular data in a cohort of 8 hyperlysinemia patients with distinct neurological features. We found novel causal mutations in AASS in all affected individuals, including 4 missense mutations, 2 deletions and 1 duplication. In two patients originating from one family, the hyperlysinemia was caused by a contiguous gene deletion syndrome affecting AASS and PTPRZ1. Hyperlysinemia is caused by mutations in AASS. As hyperlysinemia is generally considered a benign metabolic variant, the more severe neurological disease course in two patients with a contiguous deletion syndrome may be explained by the additional loss of PTPRZ1. Our findings illustrate the importance of detailed biochemical and genetic studies in any hyperlysinemia patient.
Sigma-Aldrich
Anti-AASS antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab3
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-AASS antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab1
Pricing and availability is not currently available.