Merck
Biochemistry and molecular cell biology of diabetic complications.
Nature (2001-12-14)
M Brownlee
ABSTRACT
Diabetes-specific microvascular disease is a leading cause of blindness, renal failure and nerve damage, and diabetes-accelerated atherosclerosis leads to increased risk of myocardial infarction, stroke and limb amputation. Four main molecular mechanisms have been implicated in glucose-mediated vascular damage. All seem to reflect a single hyperglycaemia-induced process of overproduction of superoxide by the mitochondrial electron-transport chain. This integrating paradigm provides a new conceptual framework for future research and drug discovery.
Sigma-Aldrich
D-Sorbitol, 99% (GC)
Pricing and availability is not currently available.