Merck
  • Improving efficiency by hybrid TiO(2) nanorods with 1,10-phenanthroline as a cathode buffer layer for inverted organic solar cells.
Improving efficiency by hybrid TiO(2) nanorods with 1,10-phenanthroline as a cathode buffer layer for inverted organic solar cells.
ACS applied materials & interfaces (2014-01-07)
Chunming Sun, Yulei Wu, Wenjun Zhang, Nianquan Jiang, Tonggang Jiu, Junfeng Fang
ABSTRACT
We reported a significant improvement in the efficiency of organic solar cells by introducing hybrid TiO2:1,10-phenanthroline as a cathode buffer layer. The devices based on polymer thieno[3,4-b]thiophene/benzodithiophene:[6,6]-phenyl C71-butyric acid methyl ester (PTB7:PC71BM) with hybrid buffer layer exhibited an average power conversion efficiency (PCE) as high as 8.02%, accounting for 20.8% enhancement compared with the TiO2 based devices. The cathode modification function of this hybrid material could also be extended to the poly(3-hexylthiophene):[6,6]-phenyl-C61-butyric acid methyl ester (P3HT:PC61BM) system. We anticipate that this study will stimulate further research on hybrid materials to achieve more efficient charge collection and device performance.
Sigma-Aldrich
1,10-Phenanthroline, ≥99%
Pricing and availability is not currently available.