Merck
  • Effect of xanthan on flavor release from thickened viscous food model systems.
Effect of xanthan on flavor release from thickened viscous food model systems.
Journal of agricultural and food chemistry (2005-04-28)
Egle Bylaite, Jens Adler-Nissen, Anne S Meyer
ABSTRACT
The influence of xanthan concentration (0, 0.02, 0.1, 0.4, and 0.8% w/w) and bulk viscosity on the release of 20 aroma compounds of different chemical classes (5 aldehydes, 4 esters, 5 ketones, 3 alcohols, and 3 terpenes) was evaluated in xanthan-thickened food model systems having different viscosities. Interactions between flavor compounds and xanthan were assessed by measuring air-liquid partition coefficients, K, of aroma compounds in pure water and in the xanthan solutions by static headspace gas chromatography. Mass transfer of aroma compounds was estimated by dynamic headspace gas chromatography. Notably, limonene and some of the esters and aldehydes exhibited decreased K values in the presence of xanthan, indicating that the release of these volatile aroma compounds was reduced due to interaction with the xanthan matrix. The degree of interaction depended on the physicochemical characteristics of the aroma compounds. A similar tendency was observed at nonequilibrium with the decreases in release rates being most pronounced for limonene, followed by the esters and aldehydes, with no effect for ketones and an apparent "salting out" effect for alcohols. The reduction in flavor release by xanthan was thus dependent on the physicochemical properties of the aroma compounds and was apparently a result of the aroma-xanthan interactions and not influenced by the viscosity of the system itself.
Sigma-Aldrich
2,3-Hexanedione, technical grade, 90%
Pricing and availability is not currently available.
Sigma-Aldrich
Menthone, mixture of isomers, ≥97.0% (GC)
Pricing and availability is not currently available.