Merck
  • Synthesis of some new pyridine-2,6-carboxamide-derived Schiff bases as potential antimicrobial agents.
Synthesis of some new pyridine-2,6-carboxamide-derived Schiff bases as potential antimicrobial agents.
Molecules (Basel, Switzerland) (2010-07-27)
Mohamed A Al-Omar, Abd El-Galil E Amr
ABSTRACT
A series of pyridine-bridged 2,6-bis-carboxamide Schiff's bases has been prepared starting from 2,6-pyridinedicarbonyl dichloride (1) and L-alanine or 2-methyl-alanine methyl ester.The coupling of acid chloride 1 with L-alanine methyl ester hydrochloride -or 2-methylalanine methyl ester hydrochloride gave the corresponding 2,6-bis-carboxamide pyridine methyl esters 2a,b.Hydrazonolysis of 2 with hydrazine hydrate afforded the corresponding bis-hydrazides 3a,b. Treatment of 3a,b with appropriate aromatic or heterocyclic aldehydes afforded the corresponding pyridine- bridged 2,6-bis-carboxamide Schiff's bases 4a-f and 5a-f, respectively. The newly synthesized compounds 2-5 were screened for their bactericidal and fungicidal activities. Many of the obtained compounds exhibited significant antimicrobial activity, comparable to streptomycin and fusidic acid, which were used as reference antibiotic drugs.
Sigma-Aldrich
2,6-Pyridinedicarbonyl dichloride, 97%
Pricing and availability is not currently available.