Merck
  • Syntheses and properties of two-dimensional, dicationic nonlinear optical chromophores based on pyrazinyl cores.
Syntheses and properties of two-dimensional, dicationic nonlinear optical chromophores based on pyrazinyl cores.
The Journal of organic chemistry (2010-11-18)
Benjamin J Coe, John Fielden, Simon P Foxon, Madeleine Helliwell, Inge Asselberghs, Koen Clays, Kurt De Mey, Bruce S Brunschwig
ABSTRACT
Six new dicationic 2D nonlinear optical (NLO) chromophores with pyrazinyl-pyridinium electron acceptors have been synthesized by nucleophilic substitutions of 2,6-dichloropyrazine with pyridyl derivatives. These compounds have been characterized as their PF(6)(-) salts by using various techniques including electronic absorption spectroscopy and cyclic voltammetry. Large red shifts in the intense, π → π* intramolecular charge-transfer (ICT) transitions on replacing -OMe with -NMe(2) substituents arise from the stronger π-electron donor ability of the latter. Each compound shows a number of redox processes which are largely irreversible. Single crystal X-ray structures have been determined for five salts, including two nitrates, all of which adopt centrosymmetric packing arrangements. Molecular first hyperpolarizabilities β have been determined by using femtosecond hyper-Rayleigh scattering at 880 and 800 nm, and depolarization studies show that the NLO responses of the symmetric species are strongly 2D, with dominant "off-diagonal" β(zyy) components. Stark (electroabsorption) spectroscopic measurements on the ICT bands afford estimated static first hyperpolarizabilities β(0). The directly and indirectly derived β values are large, and the Stark-derived β(0) response for one of the new salts is several times greater than that determined for (E)-4'-(dimethylamino)-N-methyl-4-stilbazolium hexafluorophosphate. These Stark spectroscopic studies also permit quantitative comparisons with related 2D, binuclear Ru(II) ammine complex salts.
Sigma-Aldrich
2,6-Dichloropyrazine, 98%