Merck
  • Synthesis of (2S,4S)- and (2S,4R)-5-fluoroleucine and (2S,4S)-[5,5-2H2]-5-fluoroleucine.
Synthesis of (2S,4S)- and (2S,4R)-5-fluoroleucine and (2S,4S)-[5,5-2H2]-5-fluoroleucine.
Organic & biomolecular chemistry (2004-02-11)
Jean-Damien Charrier, David S Hadfield, Peter B Hitchcock, Douglas W Young
ABSTRACT
Syntheses of (2S,4S)- and (2S,4R)-5-fluoroleucine, and, and of (2S,4S)-[5,5-(2)H(2)]-5-fluoroleucine, have been completed. The methodology allows these compounds to be prepared in sufficient quantities for incorporation by solid-state protein synthesis into strategic sites in proteins for folding studies. X-ray structures of the epimers and have been obtained and show the presence of conformational isomerism. The torsion angles between the F-C bond and the main chain are compared with values found in a mutant of the protein ubiquitin in which (2S,4S)-5-fluoroleucine replaces leucine residues 50 and 67 in the native protein.
Sigma-Aldrich
2-Mesitylenesulfonic acid dihydrate, 97%