Merck
  • Biosynthesis of polyhydroxyalkanoate copolyester containing cyclohexyl groups by Pseudomonas oleovorans.
Biosynthesis of polyhydroxyalkanoate copolyester containing cyclohexyl groups by Pseudomonas oleovorans.
International journal of biological macromolecules (2001-10-09)
D Y Kim, S B Jung, G G Choi, Y B Kim, Y H Rhee
ABSTRACT
Production of polyhydroxyalkanoates (PHAs) substituted with cyclohexyl groups by Pseudomonas oleovorans grown with 4-cyclohexylbutyric acid (4-CHB) and its mixtures with nonanoic acid (NA) was investigated. Addition of NA to medium gave rise to an increase in the total concentration of 3-hydroxy-4-cyclohexylbutyrate repeating unit in the PHAs, indicating that the bioconversion rate of 4-CHB to polyester was significantly improved by the cometabolic effect. Increasing the proportion of NA from 1.0 to 7.5 mM at a concentration of 10 mM total carbon substrate also accelerated the uptake speed of 4-CHB by the organism and resulted in an increase of the ratio of 3-hydroxynonanoate to 3-hydroxyheptanoate from 1.28 to 2.05. Differential scanning calorimetric analysis of the PHAs bearing the corresponding functional groups showed one melting transition and one glass transition temperature varying according to the composition. These results indicated that random copolyesters were obtained from the carbon substrates used in this study.
Sigma-Aldrich
Cyclohexanebutyric acid, 99%
Pricing and availability is not currently available.