Merck
Staphylococcus aureus protein A promotes immune suppression.
mBio (2013-10-03)
Scott D Kobayashi, Frank R DeLeo
ABSTRACT
Staphylococcus aureus is a prominent cause of human infections worldwide and is notorious for its ability to acquire resistance to antibiotics. Methicillin-resistant S. aureus (MRSA), in particular, is endemic in hospitals and is the most frequent cause of community-associated bacterial infections in the United States. Inasmuch as treatment options for severe MRSA infections are limited, there is need for a vaccine that protects against such infections. However, recent efforts to generate a staphylococcal vaccine have met with little success in human clinical trials. These failures are somewhat puzzling, since the vaccine antigens tested promote opsonophagocytosis in vitro and confer protection in animal infection models. One possibility is that the pathogen inhibits (and/or fails to elicit) the development of protective immunity in humans. Indeed, S. aureus produces numerous molecules that can potentially promote immune evasion, including protein A (SpA), an immunoglobulin (Ig)-binding protein present on the bacterial surface and freely secreted into the extracellular environment. SpA binds the Fc region of antibody and the Fab regions of the B-cell receptor, processes that are known to block opsonophagocytosis and cause B-cell death in vitro. In a recent study, Falugi et al. [F. Falugi, H. K. Kim, D. M. Missiakas, and O. Schneewind, mBio 4(5):e00575-13, 2013] showed that vaccination with spa mutant S. aureus strains lacking antibody Fc- and/or Fab-binding capacity protects against subsequent challenge with the USA300 epidemic strain. The findings provide strong support for the idea that SpA promotes S. aureus immune evasion in vivo and form the foundation for a new approach in our efforts to develop a vaccine that prevents severe S. aureus infections.