Merck
  • Metabolism and disposition of 2-methoxy-4-nitroaniline in male and female Harlan Sprague Dawley rats and B6C3F1/N mice.
Metabolism and disposition of 2-methoxy-4-nitroaniline in male and female Harlan Sprague Dawley rats and B6C3F1/N mice.
Xenobiotica; the fate of foreign compounds in biological systems (2012-06-26)
James M Mathews, Qiao Zhan, Amy S Etheridge, Purvi R Patel, Sherry R Black, Troy T Banks, Timothy R Fennell, Rodney W Snyder, Jason P Burgess, Stephen D Warren, Inok Surh, Suramya Waidyanatha
ABSTRACT
The disposition of 2-Methoxy-4-nitroaniline (MNA) was investigated in male and female Harlan Sprague Dawley rats and B6C3F(1)/N mice following oral, intravenous, and dermal exposure to [(14)C]MNA at 2, 15, or 150 mg/kg. Clearance of MNA was investigated in male and female rat, mouse, and human hepatocytes. MNA was cleared slowly in hepatocytes from rat (t(1/2) = 152-424 min) and human (t(1/2) = 118-403 min) but faster in mouse (t(1/2)= 70-106 min). MNA was well-absorbed in rats and mice following oral administration and eliminated chiefly in urine (rats, 75-79%; mice, 55-68%) 72 h post dosing. Less than 1% of the radioactivity remained in tissues at 72 h. MNA was poorly absorbed following dermal application in rats (5.5%) and mice (10%) over 24 h. The major pathway of metabolism of MNA was via hydroxylation of the phenyl ring to form 6-hydroxy MNA; major metabolites detected were sulfate and glucuronide conjugates of 6-hydroxy MNA. Following oral administration, the percent of total radioactivity bound in tissues bound was highest in liver (43%) and red blood cells (30%), whereas the radioactivity bound to DNA was highest in cecum (160 pmol/mg DNA).
Sigma-Aldrich
2-Methoxy-4-nitroaniline, 98%