Merck
  • Metabolism of 5 alpha-dihydroprogesterone in women and men: 3 beta- and 3 alpha-,6 alpha-dihydroxy-5 alpha-pregnan-20-ones are major urinary metabolites.
Metabolism of 5 alpha-dihydroprogesterone in women and men: 3 beta- and 3 alpha-,6 alpha-dihydroxy-5 alpha-pregnan-20-ones are major urinary metabolites.
The Journal of clinical endocrinology and metabolism (1996-10-01)
S Chantilis, R Dombroski, C H Shackleton, M L Casey, P C MacDonald
ABSTRACT
The metabolism of 5 alpha-dihydroprogesterone (5 alpha-DHP) in women and men was evaluated by defining the pattern and identity of selected metabolites excreted in urine after the iv infusion of radiolabeled 5 alpha-DHP. Virtually all of the radioactivity in urine (approximately 37% of the administered dose) was excreted within 72 h. Quantitatively, the 2 major urinary metabolites of 5 alpha-DHP in each of 13 studies conducted in 7 women and 2 men were 3 beta,6 alpha-dihydroxy-5 alpha-pregnan-20-one and 5 alpha-pregnane-3 alpha,20 alpha-diol, which could be extracted after beta-glucuronidase, but not solvolysis, treatment of the urine. Radiolabeled 3 alpha,6 alpha dihydroxy-5 alpha-pregnan-20-one (glucuronoside), in lesser amounts, also was identified in the urine of each subject. The 3 alpha/beta, 6 alpha-dihydroxy-5 alpha-pregnan-20-ones arise through specific extrahepatic pathways of progesterone/5 alpha-DHP metabolism. These metabolites are not the products of the enzyme reaction catalyzed by the cytochrome P450 steroid 6 alpha-hydroxylase of human liver (and other tissues), which affects the 6 alpha-hydroxylation of C19- and C21-delta 4-3-ketosteroids (e.g., progesterone, testosterone, and cortisol), but does not act upon 5 alpha-reduced steroids. Moreover, the steroid 5 alpha-reductases do not act upon 6 alpha-hydroxy-delta 4-3-ketosteroids. In addition, the 6 alpha-hydroxylation of 5 alpha-reduced-3 alpha/beta-hydroxysteroids is not demonstrable in adult liver tissue. Rather, the formation of 6 alpha-hydroxylated-5 alpha-pregnane-3 alpha/beta-ol-20-ones is indicative of an extrahepatic pathway of progesterone metabolism, viz. progesterone-->5 alpha-DHP-->5 alpha-pregnan-3 beta/alpha-ol-20-one(s)-->3 beta/alpha,6 alpha-dihydroxy-5 alpha-pregnan-20-one(s), in which 5 alpha-pregnan-3 alpha/beta-ol-20-ones are metabolized by an enzyme(s) that catalyzes the 6 alpha-hydroxylation of saturated substrates. There are important differences among mammalian species in the enzymes that catalyze the C-6-hydroxylation of 5 alpha-reduced C19- and C(21)-3 beta/alpha-hydroxysteroids, but in all species studied, these enzymatic reactions are the final steps in the extrahepatic inactivation of 5 alpha-reduced bioactive metabolites of progesterone (or testosterone).
Sigma-Aldrich
5α-Pregnane-3,20-dione (allo), progesterone metabolite
Pricing and availability is not currently available.