Merck
  • Simultaneous quantification of sialyloligosaccharides from human milk by capillary electrophoresis.
Simultaneous quantification of sialyloligosaccharides from human milk by capillary electrophoresis.
Analytical biochemistry (2007-09-01)
Yuanwu Bao, Libin Zhu, David S Newburg
ABSTRACT
The acidic oligosaccharides of human milk are predominantly sialyloligosaccharides. Pathogens that bind sialic acid-containing glycans on their host mucosal surfaces may be inhibited by human milk sialyloligosaccharides, but testing this hypothesis requires their reliable quantification in milk. Sialyloligosaccharides have been quantified by anion exchange high-performance liquid chromatography (HPLC), reverse- or normal-phase HPLC, and capillary electrophoresis (CE) of fluorescent derivatives; in milk, these oligosaccharides have been analyzed by high pH anion exchange chromatography with pulsed amperometric detection and, in our laboratory, by CE with detection at 205nm. The novel method described here uses a running buffer of aqueous 200mM NaH2PO4 (pH 7.05) containing 100mM sodium dodecyl sulfate (SDS) mixed with 45% (v/v) methanol to baseline resolve 5 oligosaccharides and separate all 12. This allows automated simultaneous quantification of the 12 major sialyloligosaccharides of human milk in a single 35-min run. This method revealed differences in sialyloligosaccharide concentrations between less and more mature milk from the same donors. Individual donors also varied in expression of sialyloligosaccharides in their milk. Thus, the facile quantification of sialyloligosaccharides by this method is suitable for measuring variation in expression of specific sialyloligosaccharides in milk and their relationship to decreased risk of specific diseases in infants.