Merck
  • Adverse hypersensitivity reactions to mesna as adjunctive therapy for cyclophosphamide.
Adverse hypersensitivity reactions to mesna as adjunctive therapy for cyclophosphamide.
Pediatric blood & cancer (2005-12-08)
Seong L Khaw, Peter A Downie, Keith D Waters, David M Ashley, John A Heath
ABSTRACT
Mesna is widely used for the prevention of cyclophosphamide-related hemorrhagic cystitis. It has been associated with hypersensitivity-like cutaneous and systemic reactions in adult patients. We report a series of children with malignant disease, who developed such reactions following mesna administration and discuss possible mechanisms and management issues.
Sigma-Aldrich
2-Mercaptoethanesulfonic acid solution, for protein sequence analysis, ampule, 3.0 M±0.1 M in H2O (T)
Pricing and availability is not currently available.