Merck
  • Detection of microbial contamination in fermentation processes: mass spectrometric determination of gram-negative bacteria in Leuconostoc mesenteroides cultures.
Detection of microbial contamination in fermentation processes: mass spectrometric determination of gram-negative bacteria in Leuconostoc mesenteroides cultures.
Biotechnology and bioengineering (1990-04-05)
I Elmroth, A Valeur, G Odham, L Larsson
ABSTRACT
Gas chromatography/mass spectrometry was used to detect the presence of Enterobacter cloacae in cultures of Leuconostoc mesenteroides, an organism used in an industrial process for production of dextrane. The penta-fluorobenzoyl-methyl ester derivative of 3-hydroxy-myristic acid, a characteristic compound of gram-negative bacteria, was used as the analyte. By using gas chromatography/mass spectrometry with selected ion monitoring, E. cloacae was determined over the range of 1 ppm to 1% in cultures of L. mesenteroides. The proposed analytical approach represents a useful alternative to conventional methods for determining contaminating organisms in industrial fermentation processes.
Sigma-Aldrich
2,3,4,5,6-Pentafluorobenzoyl chloride, 99%
Pricing and availability is not currently available.