Merck
[Study of plasma and hepatic hydrolysis of nitroxinil derivatives].
Annales pharmaceutiques francaises (1996-01-01)
G Baziard-Mouysset, M Alvinerie, P Galtier, M Ane-Margail, R Floch, M Payard
ABSTRACT
Seven esters derivatives of Nitroxinil were prepared and their structures were assigned by IR and 1H-NMR spectroscopy. The rate of plasma and hepatic hydrolysis were evaluated in vitro in sheep and rabbit. In view of this profile of activity, pivaloyl derivative merits evaluation, in vivo.
Product Description
Supelco
Nitroxinil, VETRANAL®, analytical standard
Pricing and availability is not currently available.