- Metabolism and degradation of betamethasone 17-valerate in homogenized living skin equivalent.
Metabolism and degradation of betamethasone 17-valerate in homogenized living skin equivalent.
Dermatology (Basel, Switzerland) (1994-01-01)
K Kubota, J Ademola, H I Maibach
PMID8305748
ABSTRACT
The metabolism of betamethasone 17-valerate was estimated using an artificial living skin equivalent (LSE). Betamethasone 17-valerate, betamethasone 21-valerate and betamethasone were measured by a normal-phase high-performance liquid chromatographic (HPLC) method. Betamethasone 17-valerate was added to the culture medium with or without LSE homogenate. Degradation profiles (%) of betamethasone 17-valerate remaining in the culture medium with skin homogenate did not differ from those without homogenate. However, the conversion of betamethasone 21-valerate to betamethasone was accelerated by skin homogenate, indicating that LSE has a sufficient level of esterase.
Product Description
Betamethasone 21-valerate, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.