Merck
  • NMR study of whole rat bile: the biliary excretion of 4-cyano-N,N-dimethyl aniline by an isolated perfused rat liver and a liver in situ.
NMR study of whole rat bile: the biliary excretion of 4-cyano-N,N-dimethyl aniline by an isolated perfused rat liver and a liver in situ.
Journal of pharmaceutical and biomedical analysis (1995-05-01)
D A Ryan, J K Sanders, G C Curtis, H Hughes
ABSTRACT
The structure of two biliary metabolites of 4-cyano-N,N-dimethyl aniline (CDA) contained in whole rat bile have been studied in detail by NMR at 400 MHz. A 4-cyano-N-methyl glutathione-N-aniline conjugate was identified as a biliary metabolite of CDA using relatively simple 1H NMR techniques. Isotopically 13C labelled CDA was used to generate 13C labelled xenobiotic conjugates. Our use of 1H/13C heteronuclear NMR techniques, in particular a 13C-selective HMQC-TOCSY experiment, allowed a N-beta-glucuronide conjugate, a previously unknown biliary metabolite of CDA, to be identified. Bile samples obtained from both the isolated perfused rat liver and the rat liver in situ were analysed.
Sigma-Aldrich
4-(Dimethylamino)benzonitrile, 98%