Merck
  • Localization of lysines acetylated in ubiquitin reacted with p-nitrophenyl acetate.
Localization of lysines acetylated in ubiquitin reacted with p-nitrophenyl acetate.
Archives of biochemistry and biophysics (1985-04-01)
J R Jabusch, H F Deutsch
ABSTRACT
The protein ubiquitin undergoes extensive N epsilon-acetylation of some of its seven lysine residues when reacted with p-nitrophenyl acetate. Lysines 27 and 29 show little reactivity whereas residue 6 is the most readily acetylated. Residues 11, 33, 48, and 63 show intermediate reactivities.
Sigma-Aldrich
Diethylmalonic acid, 98%