- Diagnostic importance of vitamin K1 and its epoxide measured in serum of dogs exposed to an anticoagulant rodenticide.
Diagnostic importance of vitamin K1 and its epoxide measured in serum of dogs exposed to an anticoagulant rodenticide.
Administration of vitamin K1, SC, to anticoagulant-poisoned (diphenadione) dogs provided diagnostic information within 4 hours, when vitamin K1 and its epoxide were measured in canine sera. Twelve dogs (2 groups of 6) were given 2.5 mg of diphenadione/kg of body weight for 3 days. Dogs were treated with vitamin K1, 2.5 (n = 6) or 5 mg/kg/day (n = 6) SC for 21 days, and their responses were compared. Four nonexposed control dogs were given 5 mg of vitamin K1/kg/day. Serum concentration of vitamin K epoxide was significantly (P less than 0.02) higher in diphenadione-exposed dogs than in control dogs 1 to 4 hours after the initial vitamin K1 treatment on day 4. Vitamin K epoxide/vitamin K1 ratios were similarly higher and became more distinct. Cessation of vitamin K1 therapy on day 24 resulted in prolongation of one-stage prothrombin times in diphenadione-exposed dogs, becoming clearly evident on day 27. Serum vitamin K1 concentrations were not detectable on day 27 in diphenadione-exposed dogs, whereas serum vitamin K1 concentrations were readily detectable in control dogs. One-stage prothrombin time changes, during days 24 to 32, indicated 5 mg of vitamin K1/kg provided better protection than did 2.5 mg of vitamin K1/kg. Coagulopathy in the dogs was resolved by day 32.