Merck
Vestibular rehabilitation with virtual reality in Ménière's disease.
Brazilian journal of otorhinolaryngology (2013-06-08)
Adriana Pontin Garcia, Mauricio Malavasi Ganança, Flávia Salvaterra Cusin, Andreza Tomaz, Fernando Freitas Ganança, Heloisa Helena Caovilla
ABSTRACT
Virtual reality technology can provide a wide range of sensory stimuli to generate conflicts of varying degrees of complexity in a safe environment. To verify the effect of a virtual realitybased balance rehabilitation program for patients with Menière's disease. This observational clinical study included 44 patients aged between 18 and 60 years diagnosed with Menière's disease submitted to a controlled randomized therapeutic intervention. The case and control groups took betahistine and followed a diet. Case group subjects underwent 12 rehabilitation sessions with virtual reality stimuli in a Balance Rehabilitation Unit (BRU TM). Patients were assessed based on DHI scores, the dizziness visual analogue scale, and underwent posturography with virtual reality before and after the intervention. After the intervention, the case group showed significantly lower scores in DHI (p < 0,001) and in the dizziness visual analog scale (p = 0.012), and had significantly greater limit of stability areas (p = 0.016) than controls. Virtual reality-based balance rehabilitation effectively improved dizziness, quality of life, and limit of stability of patients with Menière's disease.
Sigma-Aldrich
2-(2-Methylaminoethyl)pyridine, 97%
Pricing and availability is not currently available.
Betahistine mesilate, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.