- Two-dimensional copper(I) coordination polymer materials as photocatalysts for the degradation of organic dyes.
Two-dimensional copper(I) coordination polymer materials as photocatalysts for the degradation of organic dyes.
Inorganic chemistry (2012-12-19)
Tian Wen, De-Xiang Zhang, Jian Zhang
PMID23244571
ABSTRACT
Two isomeric two-dimensional copper(I) coordination polymer materials based on an in situ generated 5-(3-pyridyl)tetrazole ligand show similar layer structures but distinct photoluminescent and photocatalytic properties, which present an interesting comparative study on the structure-property correlation between isomeric materials.