Merck
  • [Pharmaceutical prescription for multiple sclerosis : evaluation of pharmaceutical consumption at private health insurance].
[Pharmaceutical prescription for multiple sclerosis : evaluation of pharmaceutical consumption at private health insurance].
Der Nervenarzt (2012-12-18)
F Wild
ABSTRACT
For persons covered by statutory health insurance (SHI) an increase in the number of defined daily doses (DDD) for pharmaceuticals to treat multiple sclerosis (MS) is known but so far there has been no comparable survey for private health insurance (PHI). Moreover, there are gaps in knowledge of the reasons for the increase and concerning the number of the MS patients in Germany. The study is based on pharmaceutical data of the PHI in Germany. The projection takes into account the different prevalence and the different male/female relationship in SHI and PHI in an extrapolation to the total population. From 2006 to 2010 the number of DDDs of MS pharmaceuticals increased by approximately 91.6 % (SHI 39.9 %) per insured person. The increase in the PHI is mainly based for on an increase in the number of MS patients. The total number of MS patients in Germany was estimated to be approximately 146,000 whereby some 12,700 MS patients (8.7 %) were insured in PHI. There is a need for research into the reasons for the increase in MS patients. The disproportional increase in the PHI compared to SHI could be a result of the increase of insured persons and an increased inclusion of persons with a higher risk of the disease.
Product Description
Interferon β-1a, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.