Merck
  • Ser1292 autophosphorylation is an indicator of LRRK2 kinase activity and contributes to the cellular effects of PD mutations.
Ser1292 autophosphorylation is an indicator of LRRK2 kinase activity and contributes to the cellular effects of PD mutations.
Science translational medicine (2012-12-18)
Zejuan Sheng, Shuo Zhang, Daisy Bustos, Tracy Kleinheinz, Claire E Le Pichon, Sara L Dominguez, Hilda O Solanoy, Jason Drummond, Xiaolin Zhang, Xiao Ding, Fang Cai, Qinghua Song, Xianting Li, Zhenyu Yue, Marcel P van der Brug, Daniel J Burdick, Janet Gunzner-Toste, Huifen Chen, Xingrong Liu, Anthony A Estrada, Zachary K Sweeney, Kimberly Scearce-Levie, John G Moffat, Donald S Kirkpatrick, Haitao Zhu
ABSTRACT
Mutations in the leucine-rich repeat kinase 2 (LRRK2) gene are the most common cause of familial Parkinson's disease (PD). Although biochemical studies have shown that certain PD mutations confer elevated kinase activity in vitro on LRRK2, there are no methods available to directly monitor LRRK2 kinase activity in vivo. We demonstrate that LRRK2 autophosphorylation on Ser(1292) occurs in vivo and is enhanced by several familial PD mutations including N1437H, R1441G/C, G2019S, and I2020T. Combining two PD mutations together further increases Ser(1292) autophosphorylation. Mutation of Ser(1292) to alanine (S1292A) ameliorates the effects of LRRK2 PD mutations on neurite outgrowth in cultured rat embryonic primary neurons. Using cell-based and pharmacodynamic assays with phosphorylated Ser(1292) as the readout, we developed a brain-penetrating LRRK2 kinase inhibitor that blocks Ser(1292) autophosphorylation in vivo and attenuates the cellular consequences of LRRK2 PD mutations in vitro. These data suggest that Ser(1292) autophosphorylation may be a useful indicator of LRRK2 kinase activity in vivo and may contribute to the cellular effects of certain PD mutations.
Sigma-Aldrich
O-Phospho-DL-serine, ≥98.0% (NT)