Merck
  • The anticholinergic and antiglutamatergic drug caramiphen reduces seizure duration in soman-exposed rats: synergism with the benzodiazepine diazepam.
The anticholinergic and antiglutamatergic drug caramiphen reduces seizure duration in soman-exposed rats: synergism with the benzodiazepine diazepam.
Toxicology and applied pharmacology (2012-02-09)
M K Schultz, L K M Wright, M F Stone, J E Schwartz, N R Kelley, M C Moffett, R B Lee, L A Lumley
ABSTRACT
Therapy of seizure activity following exposure to the nerve agent soman (GD) includes treatment with the anticonvulsant diazepam (DZP), an allosteric modulator of γ-aminobutyric acid A (GABA(A)) receptors. However, seizure activity itself causes the endocytosis of GABA(A) receptors and diminishes the inhibitory effects of GABA, thereby reducing the efficacy of DZP. Treatment with an N-methyl-d-aspartic acid (NMDA) receptor antagonist prevents this reduction in GABAergic inhibition. We examined the efficacy of the NMDA receptor antagonist caramiphen edisylate (CED; 20mg/kg, im) and DZP (10mg/kg, sc), administered both separately and in combination, at 10, 20 or 30min following seizure onset for attenuation of the deleterious effects associated with GD exposure (1.2 LD(50); 132μg/kg, sc) in rats. Outcomes evaluated were seizure duration, neuropathology, acetylcholinesterase (AChE) activity, body weight, and temperature. We also examined the use of the reversible AChE inhibitor physostigmine (PHY; 0.2mg/kg, im) as a therapy for GD exposure. We found that the combination of CED and DZP yielded a synergistic effect, shortening seizure durations and reducing neuropathology compared to DZP alone, when treatment was delayed 20-30min after seizure onset. PHY reduced the number of animals that developed seizures, protected a fraction of AChE from GD inhibition, and attenuated post-exposure body weight and temperature loss independent of CED and/or DZP treatment. We conclude that: 1) CED and DZP treatment offers considerable protection against the effects of GD and 2) PHY is a potential therapeutic option following GD exposure, albeit with a limited window of opportunity.
Sigma-Aldrich
Caramiphen hydrochloride, ≥98% (HPLC)