Merck
  • Insights into the homocoupling reaction of 4-methylamino benzoic acid mediated by Trametes versicolor laccase.
Insights into the homocoupling reaction of 4-methylamino benzoic acid mediated by Trametes versicolor laccase.
Molecular bioSystems (2011-09-14)
Andrea Martorana, Caterina Bernini, Daniela Valensin, Adalgisa Sinicropi, Rebecca Pogni, Riccardo Basosi, Maria Camilla Baratto
ABSTRACT
Spectroscopic measurements combined with Density Functional Theory calculations were applied to the characterization of the homocoupling reaction of 4-methylamino benzoic acid mediated by laccase.
Sigma-Aldrich
4-(Methylamino)benzoic acid, 97%
Pricing and availability is not currently available.