- A coincidence of addiction to "Kratom" and severe primary hypothyroidism.
A coincidence of addiction to "Kratom" and severe primary hypothyroidism.
Journal of addiction medicine (2011-08-06)
Sergey V Sheleg, Gregory B Collins
PMID21817918
ABSTRACT
Here we present a case of a coincidence of addiction to "Kratom" (botanically known as Mitragyna speciosa Korth) and developed severe primary hypothyroidism. We are discussing a possibility that high dose of indole alkaloid mitragynine (the major alkaloid identified from "Kratom") might reduce the normal response of the thyroid gland to thyroid-stimulating hormone resulting in primary hypothyroidism. Further experimental investigations of mitragynine as a possible suppressor of thyroid gland function would be a matter of interest.
Product Description
Supelco
Mitragynine solution, 100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Pricing and availability is not currently available.