Merck
  • N-methylation of the amide bond by methyltransferase asm10 in ansamitocin biosynthesis.
N-methylation of the amide bond by methyltransferase asm10 in ansamitocin biosynthesis.
Chembiochem : a European journal of chemical biology (2011-06-18)
Yingying Wu, Qianjin Kang, Guangdong Shang, Peter Spiteller, Brian Carroll, Tin-Wein Yu, Wenjin Su, Linquan Bai, Heinz G Floss
ABSTRACT
Ansamitocins are potent antitumor agents produced by Actinosynnema pretiosum. As deduced from their structures, an N-methylation on the amide bond is required among the various modifications. The protein encoded by asm10 belongs to the SAM-dependent methyltransferase family. Through gene inactivation and complementation, asm10 was proved to be responsible for the N-methylation of ansamitocins. Asm10 is a 33.0 kDa monomer, as determined by gel filtration. By using N-desmethyl-ansamitocin P-3 as substrate, the optimal temperature and pH for Asm10 catalysis were determined to be 32 °C and 10.0, respectively. Asm10 also showed broad substrate flexibility toward other N-desmethyl-ansamycins and synthetic indolin-2-ones. Through site-directed mutagenesis, Asp154 and Leu155 of Asm10 were confirmed to be essential for its catalysis, possibly through the binding of SAM. The characterization of this unique N-methyltransferase has enriched the toolbox for engineering N-methylated derivatives from both natural and synthetic compounds; this will allow known potential drugs to be modified.
Sigma-Aldrich
Ansamitocin P-3 from Actinosynnema pretiosum, ≥90% (HPLC)