Merck
  • Efficacy of taurolidine against periodontopathic species--an in vitro study.
Efficacy of taurolidine against periodontopathic species--an in vitro study.
Clinical oral investigations (2011-05-25)
Sigrun Eick, Sabrina Radakovic, Wolfgang Pfister, Sandor Nietzsche, Anton Sculean
ABSTRACT
The antimicrobial effect of taurolidine was tested against periodontopathic species in comparison to chlorhexidine digluconate in the presence or absence of serum. Minimal inhibitory concentrations (MIC), microbiocidal concentrations (MBC), as well as killing were determined against 32 different microbial strains including 3 Porphyromonas gingivalis, 3 Aggregatibacter actinomycetemcomitans, and 15 potentially superinfecting species with and without 25% v/v human serum. The MIC(50) of taurolidine against the tested microbial strains was 0.025% and the MIC(90) 0.05%. The respective values for the MBCs were 0.05% and 0.1%. Addition of 25% serum (heat-inactivated) did not change the MIC and MBC values of taurolidine. In contrast, MICs and MBCs of chlorhexidine (CHX) increased by two steps after addition of serum. Taurolidine killed microorganisms in a concentration and time-dependent manner, the killing rate of 1.6% taurolidine was 99.08% ± 2.27% in mean after 2 h. Again, killing activity of taurolidine was not affected if serum was added, whereas addition of inactivated serum clearly reduced the killing rate of all selected bacterial strains by CHX. Therefore, taurolidine possesses antimicrobial properties which are not reduced in the presence of serum as a main component in gingival crevicular fluid and wound fluid. Taurolidine may have potential as an antimicrobial agent in non-surgical and surgical periodontal treatment.
Sigma-Aldrich
Taurolidine, >97% (NMR), powder