Merck
  • Assay for beta-ureidopropionase by high-performance liquid chromatography.
Assay for beta-ureidopropionase by high-performance liquid chromatography.
Analytical biochemistry (1990-07-01)
G Waldmann, B Podschun
ABSTRACT
A sensitive assay for beta-ureidopropionase based on derivatization of the reaction product beta-alanine with phenylisothiocyanate has been developed. Purification of the resulting phenylthiocarbamoyl-beta-alanine is achieved on a LiChrospher 100 C18 reversed-phase high-performance liquid chromatography column using an isocratic elution system. Phenylthiocarbamoyl-beta-alanine is detected by its absorbance at 245 nm and quantitated by automatic peak integration referring to a calibration curve. This technique offers a high degree of sensitivity as beta-alanine quantities in the picomole range can be identified. N-Carbamoyl-beta-alanine, the natural substrate of beta-ureidopropionase, does not interfere with the described assay system. The enzymatic reaction is linear for an incubation time of 45 min with enzyme concentrations of 3.2 micrograms/ml.
Sigma-Aldrich
3-Ureidopropionic acid, ≥98.0% (T)
Pricing and availability is not currently available.