Merck
  • First evaluation of the biologic effectiveness factors of boron neutron capture therapy (BNCT) in a human colon carcinoma cell line.
First evaluation of the biologic effectiveness factors of boron neutron capture therapy (BNCT) in a human colon carcinoma cell line.
International journal of radiation oncology, biology, physics (2010-10-12)
Maria Alejandra Dagrosa, Martín Crivello, Marina Perona, Silvia Thorp, Gustavo Alberto Santa Cruz, Emiliano Pozzi, Mariana Casal, Lisa Thomasz, Romulo Cabrini, Steven Kahl, Guillermo Juan Juvenal, Mario Alberto Pisarev
ABSTRACT
DNA lesions produced by boron neutron capture therapy (BNCT) and those produced by gamma radiation in a colon carcinoma cell line were analyzed. We have also derived the relative biologic effectiveness factor (RBE) of the neutron beam of the RA-3- Argentine nuclear reactor, and the compound biologic effectiveness (CBE) values for p-boronophenylalanine ((10)BPA) and for 2,4-bis (α,β-dihydroxyethyl)-deutero-porphyrin IX ((10)BOPP). Exponentially growing human colon carcinoma cells (ARO81-1) were distributed into the following groups: (1) BPA (10 ppm (10)B) + neutrons, (2) BOPP (10 ppm (10)B) + neutrons, (3) neutrons alone, and (4) gamma rays ((60)Co source at 1 Gy/min dose-rate). Different irradiation times were used to obtain total absorbed doses between 0.3 and 5 Gy (±10%) (thermal neutrons flux = 7.5 10(9) n/cm(2) sec). The frequency of micronucleated binucleated cells and the number of micronuclei per micronucleated binucleated cells showed a dose-dependent increase until approximately 2 Gy. The response to gamma rays was significantly lower than the response to the other treatments (p < 0.05). The irradiations with neutrons alone and neutrons + BOPP showed curves that did not differ significantly from, and showed less DNA damage than, irradiation with neutrons + BPA. A decrease in the surviving fraction measured by 3-(4,5-dimetiltiazol-2-il)-2,5-difeniltetrazolium bromide (MTT) assay as a function of the absorbed dose was observed for all the treatments. The RBE and CBE factors calculated from cytokinesis block micronucleus (CBMN) and MTT assays were, respectively, the following: beam RBE: 4.4 ± 1.1 and 2.4 ± 0.6; CBE for BOPP: 8.0 ± 2.2 and 2.0 ± 1; CBE for BPA: 19.6 ± 3.7 and 3.5 ± 1.3. BNCT and gamma irradiations showed different genotoxic patterns. To our knowledge, these values represent the first experimental ones obtained for the RA-3 in a biologic model and could be useful for future experimental studies for the application of BNCT to colon carcinoma.
Sigma-Aldrich
4-Borono-L-phenylalanine, ≥95.0% (HPLC)
Pricing and availability is not currently available.