- [A new method of investigation of "child's" behavior (infant-mother attachment) of newborn rats].
[A new method of investigation of "child's" behavior (infant-mother attachment) of newborn rats].
Zhurnal vysshei nervnoi deiatelnosti imeni I P Pavlova (2010-05-18)
I S Stovolosov, V A Dubynin, A A Kamenskiĭ
PMID20469600
ABSTRACT
A new method of studying "child's" (maternal bonding) behavior of newborn rats was developed. The efficiency of the method was proved in estimation of dopaminergic control of the infant-mother attachment. Selective D2-antagonist clebopride applied in subthreshold for motor activity doses caused a decrease in aspiration of pups to be in contact with a dam. On the basis of features analyzed (latent periods and expression of various behavioral components), the integrated criterion for the estimation of "child's" reactions was suggested. Application of this criterion made it possible to neutralize high individual variability of the behavior typical of newborns.