Merck
  • Synthesis of (-)-(S,S)-clemastine by invertive N --> C aryl migration in a lithiated carbamate.
Synthesis of (-)-(S,S)-clemastine by invertive N --> C aryl migration in a lithiated carbamate.
Organic letters (2010-04-22)
Anne M Fournier, Robert A Brown, William Farnaby, Hideki Miyatake-Ondozabal, Jonathan Clayden
ABSTRACT
The first enantioselective synthesis of the antihistamine agent clemastine, as its (S,S)-stereoisomer, has been achieved by ether formation between a proline-derived chloroethylpyrrolidine and an enantiomerically enriched tertiary alcohol. The tertiary alcohol was formed from the carbamate derivative of alpha-methyl-p-chlorobenzyl alcohol by invertive aryl migration on lithiation. The (S,S)-stereochemistry of the product confirms the invertive nature of the rearrangement.
Sigma-Aldrich
(S)-4-Chloro-α-methylbenzyl alcohol, 95%
Sigma-Aldrich
Clemastine fumarate salt, ≥98% (HPLC)