Merck
  • Oral uridine supplementation antagonizes the peripheral neuropathy and encephalopathy induced by antiretroviral nucleoside analogues.
Oral uridine supplementation antagonizes the peripheral neuropathy and encephalopathy induced by antiretroviral nucleoside analogues.
AIDS (London, England) (2009-12-25)
Nils Venhoff, Dirk Lebrecht, Catherine Deveaud, Bertrand Beauvoit, Jacques Bonnet, Klaus Müller, Janbernd Kirschner, Ana C Venhoff, Ulrich A Walker
ABSTRACT
Peripheral neuropathy and central nervous system neurodegeneration may result from the mitochondrial toxicity of some antiretroviral nucleoside analogues. We investigated whether this neuropathology may be antagonized by uridine supplementation in vivo. Because of the obvious difficulties in obtaining human neural tissues, the mitochondrial neurotoxicity of the nucleoside analogues was studied in mice. BALB/C mice (7 weeks of age) were fed for 9 weeks with zalcitabine (13 mg/kg per day) or zidovudine (100 mg/kg per day) with or without mitocnol (340 mg/kg per day), a dietary supplement with high uridine bioavailability. Hippocampal and sciatic nerve mitochondria were analyzed. Zalcitabine and to a lesser extent zidovudine induced a significant peripheral neuropathy and encephalopathy with disrupted mitochondrial ultrastructure, depleted mitochondrial DNA, reduced levels of cytochrome c oxidase activity and diminished expression of mitochondrial DNA-encoded cytochrome c oxidase subunit I. Mitocnol had no intrinsic effects but attenuated or fully normalized all measured disorder of the peripheral and central nervous system. Zidovudine and zalcitabine induce a mitochondrial disorder in the peripheral and central nervous system, both of which are antagonized by uridine supplementation.
Sigma-Aldrich
2′,3′-Dideoxycytidine, ≥98% (HPLC)