Merck
  • Matrix solid-phase dispersion and solid-phase microextraction applied to study the distribution of fenbutatin oxide in grapes and white wine.
Matrix solid-phase dispersion and solid-phase microextraction applied to study the distribution of fenbutatin oxide in grapes and white wine.
Analytical and bioanalytical chemistry (2009-10-07)
R Montes, P Canosa, J Pablo Lamas, A Piñeiro, I Orriols, R Cela, I Rodríguez
ABSTRACT
The fate of the acaricide fenbutatin oxide (FBTO) during the elaboration of white wine is evaluated. Matrix solid-phase dispersion (MSPD) and solid-phase microextraction (SPME) were used as sample preparation techniques applied to the semi-solid and the liquid matrices involved in this research, respectively. Selective determination of FBTO was achieved by gas chromatography with atomic emission detection (GC-AED). GC coupled to mass spectrometry was also used to establish the identity of FBTO by-products detected in must and wine samples. MSPD extractions were accomplished using C18 as dispersant and co-sorbent. Sugars and other polar interferences were first removed with water and water/acetone mixtures, then FBTO was recovered with 8 mL of acetone. When used in combination with GC-AED, the MSPD method provided limits of quantification (LOQs) in the low nanogram per gram range, recoveries around 90% and relative standard deviations below 13% for extractions performed in different days. Performance of SPME for must and wine was mainly controlled by the extraction temperature, time and fibre coating. Under final conditions, FBTO was extracted in the headspace mode for 45 min at 100 degrees C, using a 100 microm poly(dimethylsiloxane)-coated fibre. The achieved LOQs remained around or below 0.1 ng mL(-1), depending on the type of sample, and the inter-day precision ranged from 10% to 13%. FBTO residues in grapes stayed mostly on the skin of the fruit. Although FBTO was not removed during must and white wine elaboration, it remained associated with suspended particles existing in must and lees, settled after must fermentation, with a negligible risk of being transferred to commercialised wine. On the other hand, two by-products of FBTO (bis and mono (2-methyl-2-phenylpropyl) tin) were identified, for first time, in must and final white wines obtained from FBTO treated grapes. Found values for the first species ranged from 0.03 to 0.9 ng mL(-1).
Product Description
Supelco
Fenbutatin oxide, PESTANAL®, analytical standard
Pricing and availability is not currently available.