Merck
  • Effect of intracanal corticosteroids on healing of replanted dog teeth after extended dry times.
Effect of intracanal corticosteroids on healing of replanted dog teeth after extended dry times.
Journal of endodontics (2009-05-05)
Anna Kirakozova, Fabricio B Teixeira, Alice E Curran, Fang Gu, Peter Z Tawil, Martin Trope
ABSTRACT
This study investigated the effect of potent intracanal corticosteroids on periodontal healing of replanted avulsed teeth and evaluated the systemic absorption of these corticosteroids. Sixty-seven extracted dog premolar roots were randomly assigned to one of the following groups: groups 1-3 filled with gutta-percha and replanted immediately and after 40 and 60 minutes, respectively; groups 4 and 5 filled with 0.05% clobetasol; and groups 6 and 7 filled with 0.05 % fluocinonide. Groups 4 and 6 were replanted after 40 minutes and groups 5 and 7 after 60 minutes. After 4 months, roots were evaluated histologically for signs of periodontal healing. Roots treated with clobetasol and fluocinonide healed more favorably than roots filled with gutta-percha and were different from each other at 60 minutes. No change in the systemic corticosteroid blood concentration was observed in any group. Corticosteroids were efficacious in the beagle model as intracanal medicaments for promoting favorable postavulsion periodontal healing.
Sigma-Aldrich
Fluocinonide, ≥98% (HPLC)
Pricing and availability is not currently available.