Merck
[Renal effect of treatment for heart failure].
Ugeskrift for laeger (2009-03-05)
Zhiying Zhang Rasmussen, Toke Barfod, Jakob Klit
ABSTRACT
The case of a 66-year-old male with heart failure and cardiorenal syndrome is presented. The patient had normal renal function before intensive treatment with diuretics and ACE inhibitor. Shortly after the ACE inhibitor was stopped and diuretics were either stopped or reduced in dosage, his renal function normalized. Suggestions are presented for follow-up after initiation of ACE inhibitor treatment.