Merck
  • The syntheses and properties of tricyclic pyrrolo[2,3-d]pyrimidine analogues of S6-methylthioguanine and O6-methylguanine.
The syntheses and properties of tricyclic pyrrolo[2,3-d]pyrimidine analogues of S6-methylthioguanine and O6-methylguanine.
Organic & biomolecular chemistry (2006-04-25)
Ana R Hornillo-Araujo, Adam J M Burrell, Miren K Aiertza, Takayuki Shibata, David M Hammond, Dolorès Edmont, Harry Adams, Geoffrey P Margison, David M Williams
ABSTRACT
The syntheses of novel tricyclic pyrrolo[2,3-d]pyrimidine analogues of S6-methylthioguanine are described. The crystal structures and pKa values of these and related O6-methylguanine analogues are reported. All compounds display higher pKa values than O6-methylguanine with the sulfur-containing analogues being the more basic and exhibiting higher stability in aqueous solution. In a standard substrate assay with the human repair protein O6-methylguanine-DNA methyltransferase (MGMT) only the oxygen-containing analogue displayed activity.
Sigma-Aldrich
2-Amino-6-methylmercaptopurine, ≥95%
Pricing and availability is not currently available.