Merck
  • Influence of inorganic phosphate and organic buffers on cephalosporin production by Streptomyces clavuligerus.
Influence of inorganic phosphate and organic buffers on cephalosporin production by Streptomyces clavuligerus.
Archives of microbiology (1977-11-18)
Y Aharonowitz, A L Demain
PMID23086
ABSTRACT
A high concentration of potassium phosphate (75--100 mM) stabilized pH and supported extensive growth of Streptomyces clavuligerus in a chemically defined medium; such a concentration also inhibited cephalosporin production. Although Tris buffer was found to have detrimental effects on growth and antibiotic production, 3-(N-morpholine)-propane sulfonate (MOPS) or 2-(NP-morpholine)-ethane sulfonate (MES) buffer provided a nontoxic buffering system. In the presence of MOPS buffer, cephalosporin production was optimal at 25 mM phosphate, whereas higher concentrations of phosphate progressively inhibited antibiotic production up to 85% without modifying the pH pattern. MOPS buffer can be used to conduct fermentations at a relatively constant pH value in shake flasks.