Merck
  • Enzymatic synthesis and 19F NMR studies of 2-fluoroadenine-substituted RNA.
Enzymatic synthesis and 19F NMR studies of 2-fluoroadenine-substituted RNA.
Journal of the American Chemical Society (2004-09-24)
Lincoln G Scott, Bernhard H Geierstanger, James R Williamson, Mirko Hennig
ABSTRACT
The production of isotopically labeled RNA remains critical to current NMR structural studies. One approach to obtain simple NMR spectra is to label with a nucleus that is not naturally occurring in RNA. Fluorine-19 can serve as a sensitive site-specific probe upon incorporation into RNA. Here we report the efficient in vitro enzymatic synthesis of 2-fluoroadenosine-5'-triphosphate and its incorporation into the HIV-2 transactivation region (TAR) of RNA by DNA template-directed transcription using phage T7 RNA polymerase. We provide unequivocal evidence for this 19F-substituted base analogue capability to selectively interact with uracil, forming 2F-A-U base pairs in RNA. The introduction of a 2-fluoroadenyl substitution is relatively nonperturbing and provides us with uniquely positioned, sensitive NMR reporter groups to monitor structural changes in the local RNA environment.
Sigma-Aldrich
2-Fluoroadenine, 96%
Pricing and availability is not currently available.